Microbiology manifold
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
516-1038EA 25520 DKK
516-1038
Microbiology manifold
Manifolder
This microbiology manifold, with modular design, allows you to optimise testing without sacrificing cleanliness.
  • Can be customised to fit workflow, test area and autoclave
  • Slip-fit adapters enable the use of any Pall microbiology device on the same manifold
  • Right- or left-handed operation
With a 3-place manifold and a coupling device, an additional manifold can be added to make a 6-place manifold. A preventative maintenance schedule can be designed without delays to critical testing.
Adapters are required and need to be ordered separately.
MicroFunnel™ adapter: Ideal for MicroFunnel™, MicroFunnel™ ST, MicroFunnel™ Plus and MicroCheck® devices.
Sentino® funnel adapter: Ideal for Sentino® filter funnels.
Standard adapter: Ideal for Sentino® magnetic filter funnels.
Elongated standard adapter: Ideal for magnetic filter funnels.
Bestillingsoplysninger: The microbiology manifold is supplied as a set and comes with 3-place manifold base, 3 manifold valves, 1 end cap and 1 hose barb cap. Different adapters and coupling devices are available. Adapters are required and need to be ordered separately.