VWR®, Erlenmeyer Flasks, Narrow Neck, Heavy-Duty
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
214-0421EA 506 DKK
214-0421 214-0422 214-0423 214-0424 214-0425
VWR®, Erlenmeyer Flasks, Narrow Neck, Heavy-Duty
Kolber Erlenmeyer-kolber
Borosilicate 3.3 glass.
  • Resistant to chemicals and thermal shock
  • Volume graduations and labelling area
Heavy-duty flasks have better mechanical stability than standard products due to thicker walls. The reinforced rims increase shock resistance and reduce the risk of breakage. Due to the increased wall thickness, resistance to rapid temperature changes is not as good as those of the standard products, therefore, slow and uniform heating is recommended.