VWR® ML3 LED Illumination for Microscopes
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
631-1780EA 3860 DKK
631-1780
VWR® ML3 LED Illumination for Microscopes
Lyskilder LED-belysnings
This state-of-the-art lighting module is designed as a plug and play spare part for microscopes like the Zeiss Standard 18. The 3 W LED, with a colour temperature of 6500 K, has an average lifespan of approximately 50000 hours.
  • Aluminium housing
  • Light can be continuously dimmed
  • Last dimming position can be stored with memory function for the next use
Leveringsinformation: Supplied with a multi-adapter power supply (EU, US, UK).