DCPIP assay reagent
Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CAYM700942-700EA 536 DKK
CAYM700942-700
DCPIP assay reagent
Assays ELISA'er
The vial contains 700 μg of 2,6-dichlorophenolindophenol (DCPIP) assay reagent dissolved in UltraPure water. Thaw the reagent prior to use in the MitoCheck® Complex II Activity Assay Kit.