Precision balances, LG series
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
611-3475EA 8370 DKK
VWRI611-3475
Precision balances, LG series
LG series precision balances have a database and many embedded advanced features which are ideal for use in laboratory, research departments, formulation and quality control applications. All models have a stainless steel top pan.
  • High resolution load cell weighing system
  • Large graphic display allows efficient and easy operation
  • GLP/ISO compliant records of weight values with user ID, date, time and serial number
  • Round glass windshield and protective plastic cover
  • Under hook weighing is applicable for external calibration models
User can choose three options for models with internal calibration (denoted by "i" at end of the model name):
Balance will calibrate using the internal weight by pressing CAL button
Balance will automatically recalibrate when ambient temperature changes and/or after a certain time has passed (Incal+)
The user can choose to calibrate the balance with an external weight
Certificeringer: EN 61326-1:2013, EN 61010-1: 2010, EN 61010-2-010:2014, EN 50581:2012
Leveringsinformation: Supplied with plastic in use protective cover.
Advarsel: Models with a large graphic display are denoted by G in the model name. Similarly i shows internal calibration.