Full face mask, Promask
Leverandør: Scott Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Promask
SCTS5512681EA 1440 DKK
SCTS5512681
Full face mask, Promask
Åndedrætsværn Genanvendelige masker Fuldtdækkende ansigtsmasker
The Promask is a single filter full face mask that includes T-bar faceseal design, which provides a highly efficient seal with minimal pressure on the face. Promask incorporates a wide panoramic visor to maximise the field of vision and a front mounted speech diaphragm for easy communication with colleagues.
  • Procomp®, a durable halo-butyl elastomer provides durability and high chemical resistance
  • Clear inner mask increases wearer acceptability
  • Side (left) fitting filter port
  • Suitable for CBRN applications
An optional hard coated visor is available to enhance resistance to scratching and solvents.
Certificeringer: EN 136