Bump cap, Bumpmaster
Leverandør: Scott Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Bumpmaster
SCTS2011420EA 0 DKK
SCTS2011420
Bump cap, Bumpmaster
Hjelme Bump Caps
Bumpmaster is a lightweight and wipe clean low profile PE bump cap.
  • Comfortable protection with foam sweatband option
  • Compatable with ear defenders, spectacles and goggles
Ideally suited to applications where cleanliness in conjunction with impact protection are the prime considerations, for example, in the food preparation industry.
Certificeringer: EN 812