Surface disinfectant wipes, PURELL® Antimicrobial Wipes Plus
Leverandør: GOJO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PURELL®
GOJO9213-06-EEU00EA 625 DKK
GOJO9213-06-EEU00
Surface disinfectant wipes, PURELL® Antimicrobial Wipes Plus
Papirservietter Engangsservietter og - håndklæder Desinficerende servietter
These wipes offer outstanding efficacy against bacteria and yeast, and are active on viruses. They can be used on almost all common surfaces, such as phones, gym equipment, shopping trolleys, door handles and lift buttons. Ideal for grocery stores, offices, restaurants, health clubs or hotels.
  • Active against Coronavirus according to EN 14476
  • Broad spectrum activity (bacteria, yeast, selected viruses)
  • Easy-to-use
Advarsel: Use biocides safely. Always read the label and product information before use.
*Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904