Handheld refractometers, OPTi
Leverandør: Bellingham + Stanley
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
635-0607EA 2990 DKK
BELL38-51 BELL38-52 BELL38-53 BELL38-56
Handheld refractometers, OPTi
  • Shallow prism dish made of 316 stainless steel
  • Zero calibration with distilled water
  • Automated temperature compensation (ATC)
Leveringsinformation: Supplied with two batteries, multi-language guide, certificate of calibration and protective case.