Cell separation solution, Biocoll
Leverandør: Biochrom
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L6155EA 1590 DKK
BCHRL6155
Cell separation solution, Biocoll
Biocoll separating solution contains Ficoll 400® (Polysucrose 400), a polymer with a molecular weight of ~40 0000 Da. Densities of up to 1,1 g/ml can be adjusted using this hydrophilic polymer. For optimal pH and osmolality, adjusting Biocoll with an acid, preferably amidotrizoeic acid (ATA) and sodium hydroxide is required. Biocoll with densities of 1,077 g/ml and 1,090 g/ml are already adjusted accordingly.