Nut driver
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
REST21076EA 273 DKK
REST21076
Nut driver
møtrikdrivere
This nut driver is ideal for changing Agilent 5890/6890/6850 FID jets and securely grips jet for easy removal or installation.