Multipacks, mLINE®
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
mLINE®
613-2809EA 6920 DKK
613-2809
Multipacks, mLINE®
Pipetter Automatiske pipetter
These Multipacks combine a set of mLINE® mechanical pipettes with matching tip trays and useful accessories. mLINE® is recommended by health and safety officers around the globe. It covers the full volume range of 0,1 µl to 10 ml and is available in single- and multichannel models.
  • Excellent ergonomics to protect from injuries with the exceptionnally light pipetting and tip ejection forces
  • High accuracy and precision, also in repetitive, long-lasting pipetting
  • Minimized risk of contamination with Safe-Cone Filters and full autoclavability
  • Increased safety with volume lock preventing accidental volume changes while pipetting
It is particularly designed for repetitive and long-lasting pipetting in order to prevent work related hand, arm and shoulder disorders or Repetitive Strain Injury (RSI).
Bestillingsoplysninger: All tip racks contain 96 tips except volume 100 - 5000 µl which contains 50 tips.