Universal container, Sterilin™
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SLIN30APPRNEA 1510 DKK
SLIN30APPRN SLIN30APPRN/IRR SLIN30BPPRN SLIN30BPPRN/IRR
Universal container, Sterilin™
Quality assured containers, ideal for the exacting requirements of molecular biology and genomic research applications.
  • Free from RNase, DNase, human DNA and Pyrogens
  • ½ turn Quickstart cap for ease of handling
  • Excellent leak free performance – tested in accordance with BS EN 14254 Annexe D
  • 95kPa compliant at ambient temperature
  • Convenient self-standing conical base
  • Lot number on each container for traceability
  • Aseptically manufactured using controlled, automated cleanroom processes
  • Maximum rcf 9500 xg
Manufactured under controlled cleanroom conditions, this clear robust polypropylene container has a wealth of features which make it a laboratory essential. With a working volume of 25 ml and convenient self-standing conical base, it’s perfect for a wide range of general laboratory uses from sample handling to centrifugation applications.
Note: For research use only. Not for diagnostic applications.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904