Quality assured containers, ideal for the exacting requirements of molecular biology and genomic research applications.
- Free from RNase, DNase, human DNA and Pyrogens
- ½ turn Quickstart cap for ease of handling
- Excellent leak free performance – tested in accordance with BS EN 14254 Annexe D
- 95kPa compliant at ambient temperature
- Convenient self-standing conical base
- Lot number on each container for traceability
- Aseptically manufactured using controlled, automated cleanroom processes
- Maximum rcf 9500 xg
Manufactured under controlled cleanroom conditions, this clear robust polypropylene container has a wealth of features which make it a laboratory essential. With a working volume of 25 ml and convenient self-standing conical base, it’s perfect for a wide range of general laboratory uses from sample handling to centrifugation applications.
Note: For research use only. Not for diagnostic applications.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904