MERC1.02480
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.02480.0500EA 400 DKK
MERC1.02480.0500 MERC1.02480.1000
MERC1.02480
For a more intensive and broader counterstaining range in H&E.
  • Eosin and Phloxine bind to the positively charged plasma proteins
  • Combination of both dyes results in a broader color range in the tissue
This Eosin-Phloxine solution, alcoholic is used in Hemytoxylin and eosin (H&E) overview staining that is routinely performed in histological laboratories. Eosin and Phloxine bind to the positively charged plasma proteins. The combination of both dyes results in a broader color range in the tissue. The solution is registered as IVD and CE product and results in approx. 1500 stains per L.