Safety helmets, Tuffmaster II
Leverandør: Scott Safety
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Tuffmaster II
SCTS2011101EA 149 DKK
SCTS2011101
Safety helmets, Tuffmaster II
Hjelme Sikkerhedshjelme
Tuffmaster II offers a traditional styled head protection with ease of connectivity to face and hearing protection. Features the proven head harness in combination with a high quality robust ABS shell.
  • Perfectly balanced helmet for full shift wearing
  • Approved with the HXSPEC Eyeshield
  • Pin lock head size can be adjusted from 50 to 66 cm
  • Wheel ratchet head size can be adjusted from 50 to 64 cm
Tuffmaster II HC710: Includes soft weave 25 mm terylene headgear, standard pin lock size adjustment and a terry towelling sweatband.
Certificeringer: EN 397