Disinfectant wipes, Bactinyl®
Leverandør: Orapi
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Bactinyl®
115-2080EA 81.3 DKK
115-2080 115-2115 115-2122 115-2082
Disinfectant wipes, Bactinyl®
Papirservietter Engangsservietter og - håndklæder Desinficerende servietter
Pre-saturated wipes for cleaning, pre-disinfection and disinfection of contact equipment, sensors and instruments. The wipes kill 99,999% of all germs targeted.
  • Available perfumed or odourless
  • Can be used on all types of materials
  • Practical tub dispenser with lid
Bactericidal: NF EN 1040, NF EN 1276.
Fungicide: NF EN 1275, NF EN 1650.
Virucidal: NF T 72-180 on Rotavirus, HIV-1, Poliovirus, Herpes virus. NF EN 14476 on H1N1.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904