DNA (Deoxyribonucleic acid) Blocking agent, sheared and denatured
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E213-5MLEA 768 DKK
E213-5ML
DNA (Deoxyribonucleic acid) Blocking agent, sheared and denatured
DNA (Deoxyribonucleic acid)
An effective blocking agent used to increase the signal-to-noise ratio for detection during hybridisation.
  • Biotechnology grade
  • Can be stored in freezer
MDL Number: MFCD00130922
CAS-nummer: 9007-49-2