Tubing assemblies, single-use, C-Flex®
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
734-2840EA 2780 DKK
VWRI734-2840 VWRI734-2838 VWRI734-2839 VWRI734-2834 VWRI734-2835 VWRI734-2836 VWRI734-2837 VWRI734-2841 VWRI734-2842 VWRI734-2843 VWRI734-2832 VWRI734-2833
Tubing assemblies, single-use, C-Flex®
Unique custom designed solutions to fit for specific requirements with a broad spectrum of applications including life sciences and general lab use, environmental assays, fluid handling in single-use biopharmaceutical bags, cell culture transfer, pumping applications and medical device components.
  • Tubing is both weldable and sealable
  • Versatile and significant cost savings
  • Pre-sterilized and ready-to-use
  • Gamma irradiated to Sterility Assurance Level (SAL) 10-6
  • Tubing assemblies are offered with custom size and configuration capabilities
Products are animal free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from −20 to 65 °C without losing integrity.
Certificeringer: Manufactured from USP Class VI materials in ISO Class 7 Cleanroom.
Emballering: All assemblies are individually double bagged with product label located on inner bag.