Bulk adsorbents, granulated activated copper
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
REST26136EA 1130 DKK
REST26136
Bulk adsorbents, granulated activated copper
Kromatografi sorbenter SPE Sorbenter
This activated copper effectively removes elemental sulphur from environmental extracts. The 30 mesh granular material eliminates the potential for fine copper particles in filtered extracts.
  • Convenient form for removing sulphur from environmental extracts
  • Acidified and activated - ready for use