Bulk adsorbents, sodium sulfate
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
REST26204EA 1910 DKK
REST26204
Bulk adsorbents, sodium sulfate
Kromatografi sorbenter SPE Sorbenter
Anhydrous sodium sulphate is the most common drying agent used to remove moisture from sample extracts. Activate by heating to 400 °C for four hours before use.
Emballering: Packaged in recloseable 5 kg buckets.