Anti-IVL Mouse Monoclonal Antibody (HRP) [clone: SY5]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNCH0678-500EA 3670 DKK
BNCH0678-500 BNCH0678-100
Anti-IVL Mouse Monoclonal Antibody (HRP) [clone: SY5]
This antibody recognizes a protein of 66 kDa-170 kDa, identified as involucrin. In Western blotting of cultured human keratinocytes, this MAb reacts with a 120 kDa protein. It stains the involucrin in a variety of sizes: 170 kDa in MCF-7 cells, a doublet of ~115 kDa and 150 kDa in gorilla and owl monkey, 66 kDa in dog, and a doublet of 105 kDa in pig. Its epitope maps between codon 421-568 of human involucrin. Involucrin is expressed in a range of stratified squamous epithelia, including the cornea, which lacks a distinct cornified layer. In normal epidermis, it is first expressed in the upper spinous layers, and in keratinocyte cultures, all cells that have left the basal layer express it. Involucrin expression is altered in pathological conditions: in psoriasis and other benign epidermal hyperplasias, involucrin expression begins closer to the basal layer than normal; expression is abnormal in squamous cell carcinomas and premalignant lesions, and is reduced in severe dysplasias of the larynx and cervix.

Type: Primary
Antigen: IVL
Clonality: Monoclonal
Clone: SY5
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Dog, Gorilla, Human, Monkey, Owl, Pig
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR