Trimethylamine-N-oxide, white crystalline powder
Leverandør: MP Biomedicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
TMANO
ICNA0521947010EA
1720
DKK
ICNA0521947010
Trimethylamine-N-oxide, white crystalline powder
Trimethylamine-N-oxide
Trimethylamine N-oxide is used as demetallation and decarbonylation reagent for organometallic compounds. Mediates the conversion of thiols to disulfides.
Advarsel: Research Use Only (RUO).
Formel:
C₃H₉NO Molekylvægt: 75,11 g/mol Smeltepunkt: 220…222 °C (lit.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002048 CAS-nummer: 1184-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...