Labels for BMP™ 51/53 printer, type B-488
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WorkHorse
BRDY143308EA 634 DKK
BRDY143308 BRDY143307
Labels for BMP™ 51/53 printer, type B-488
Etiketter
Printable, high-performance polyester labels for BMP™ 41 and BMP™ 51/53 label makers.
  • Resists short term xylene exposure, DMSO and ethanol
  • Can be used for basic slide identification