Labels for BMP™ 51/53 printer, type B-499
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BRDY143343EA 449 DKK
BRDY143343 BRDY143354 BRDY143342 BRDY143353 BRDY143341 BRDY143352 BRDY143362 BRDY143340 BRDY143351 BRDY143350 BRDY143361 BRDY143360 BRDY143349 BRDY143348 BRDY143357 BRDY143346 BRDY143356 BRDY143355 BRDY143344 BRDY131588 BRDY131600
Labels for BMP™ 51/53 printer, type B-499
Etiketter
These labels featured nylon cloth with permanent adhesive that allows labelling in environments with heat, cold, oil and dirt. They are ideal for wire and cable marking, electronic component marking, laboratory vial identification and general labelling.
  • Long and short-tail styles provide window for viewing liquid level in tube
  • Thin construction prevents jamming in centrifuges and racks
  • Permanent acrylic adhesive