Labels for BMP™ 51/53 printer, type B-461
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BRDY143288EA 428 DKK
BRDY143288 BRDY143299 BRDY143287 BRDY143297 BRDY143286 BRDY143285 BRDY143284 BRDY143294 BRDY143293 BRDY143292 BRDY143291 BRDY143289
Labels for BMP™ 51/53 printer, type B-461
Etiketter
Ideal for labelling cryo, centrifuge and PCR tubes that will be exposed to extreme temperatures in the lab and hold-up against common laboratory chemicals and fit the most popular tube sizes.
  • Printable high-performance laboratory self-laminating polyester
  • Ultra-thin construction prevents jamming in racks and centrifuges
  • Suitable for use in applications with liquid nitrogen