Labels for BMP™ 71 printer, type B-481
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
StainerBondz™
BRDY710766EA 298 DKK
BRDY710766
Labels for BMP™ 71 printer, type B-481
Etiketter
These StainerBondz™ polyester labels remain resistant when exposed to xylene and other chemicals that are commonly used in the slide staining process.
  • No print removal
  • No label buckling
  • No discolouration that would cause barcodes to be unreadable
  • Stays adhered to the slide