Dow Corning® SYLGARD® 186 silicone elastomers, 66700 cP
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
634175UEA
6560
DKK
634175U
Dow Corning® SYLGARD® 186 silicone elastomers, 66700 cP
Dow Corning® SYLGARD® 186 Silicone Elastomers
Dow Corning® silicone encapsulants are supplied as two-part liquid component kits. When liquid components are thoroughly mixed, the mixture cures to a flexible elastomer, which is well suited for the protection of electrical/electronic applications.
- Rapid, versatile cure processing controlled by temperature
- Can be considered for uses requiring UL requirements
- 10:1 mix ratio with high viscosity and high tear strength