VWR®, Safety Waste Caps
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
590-0045EA 944 DKK
590-0045 590-0046 590-0043 590-0044
VWR®, Safety Waste Caps
Låg Sikkerhedslåg og sikkerhedsaffaldslåg
Collect your solvent waste safely without being exposed to solvent vapours or hazardous gases. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
  • Recommended for HPLC systems
  • Chemically resistant against aggressive organic solvents
  • Tubing connector for e.g. leak connection of diverse HPLC instruments (only for safety waste caps with tube connections)
Bestillingsoplysninger: Exhaust air filter needs to be ordered separately.