Bottle top vacuum filtration systems, GL 45, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
215-3891EA 115 DKK
215-3891
Bottle top vacuum filtration systems, GL 45, DURAN®
Filtre Topfiltre til flasker
Designed for sterilisation or clarification of aqueous cell culture media. Supplied as a filter funnel only unit for use with the DURAN® TILT bottle (with GL 45 thread adaptor) or 45 mm media bottles.
  • Comes in three different asymmetric pore sizes (0,1 μm, 0,2 μm, or 0,45 μm)
  • Raised moulded graduation marks for easy volume reading
  • Manufactured in a Class 100,000 clean room from Class VI, non-cytotoxic materials