Cryoprotective freezing medium, BioWhittaker™
Leverandør: Lonza
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
733-1686EA 1010 DKK
LONZ12-132A
Cryoprotective freezing medium, BioWhittaker™
Cryoprotective medium containing BME with Hank’s Balanced Salts Solution and 15% DMSO, without L-glutamine. Used 1:1 with growth medium containing 10% FBS.