VWR® Dropping Funnels, Cylincrical, with Pressure Balance
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
201-1250EA 754 DKK
201-1250 201-1249 201-1245 201-1246 201-1247 201-1248
VWR® Dropping Funnels, Cylincrical, with Pressure Balance
Tragte Dråbetragte
Borosilicate 3.3 glass.
  • Standard tapered joints top and bottom, drip stem and pressure-equalising arm
  • PTFE key stopcock and plastic stopper
  • Very good chemical resistance
  • Very good temperature resistance
Drip stem does not extend beyond joint, thereby eliminating a possible cause of breakage. PTFE key for grease-free performance.