Atomic absorption spectrocopy (AAS) standards, cobalt in oil, Certipur®, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.15061.0100EA 2080 DKK
MERC1.15061.0100
Atomic absorption spectrocopy (AAS) standards, cobalt in oil, Certipur®, Supelco®
Standard dissolved in oil is ready-to-use calibration standards for the analysis of cobalt by AAS in a non-aqueous matrix.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022