Alkylation reagents
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HYPETS-49350EA 4570 DKK
HYPETS-49350
Alkylation reagents
Kromatografi sorbenter GC Sorbenter
Methylate Reagent (DMFDMA) reactions are typically complete upon dissolution (except for long-chain solid acids) and quantitative yields are obtained when reagent and sample are injected without prior mixing.
  • Ready-to-use reagent
  • For easy and effective preparation of methyl esters from fatty acids and amino acids
  • No water washing, extraction or concentration of derivatives required
Emballering: Packaged in hypo-vial sample storage vial.