2-Quinoxalinol 98%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

ACRO454460250EA 400 DKK
ACRO454460250
2-Quinoxalinol 98%
2-Quinoxalinol
Formel: C₈H₆N₂O
Molekylvægt: 146,15 g/mol
Smeltepunkt: 271…272 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006722
CAS-nummer: 1196-57-2

Bestil nu

Specifikationer Testresultater

Appearance White to yellow to tan Crystalline powder
Infrared spectrum Conforms
Melting point 269 to 273 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR