1-Boc-azetidine-3-ylmethanol 96%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

ACRO452090050EA 3090 DKK
ACRO452090050
1-Boc-azetidine-3-ylmethanol 96%
1-Boc-azetidine-3-ylmethanol
Formel: C₉H₁₇NO₃
Molekylvægt: 187,24 g/mol
Smeltepunkt: 50…53 °C
Flammepunkt: 117 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06656141
CAS-nummer: 142253-56-3

Bestil nu

Specifikationer Testresultater

Appearance Colorless to white to yellow Crystals or low melting solid
Infrared spectrum Conforms
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR