(4-Bromophenyl)methanesulphonyl chloride 95%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
ACRO451490050EA
2490
DKK
ACRO451490050
(4-Bromophenyl)methanesulphonyl chloride 95%
(4-Bromophenyl)methanesulphonyl chloride
Formel:
C₇H₆BrClO₂S Molekylvægt: 269,5433 g/mol Smeltepunkt: 116…118 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04973454 CAS-nummer: 53531-69-4 UN nummer: 3261 ADR: 8,II |
Specifikationer Testresultater
Appearance | White to off-white Powder or solid |
Infrared spectrum | Conforms |
Melting point | 116 to 121 °C |
HPLC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...