5-Bromo-1,3,4-thiadiazol-2-amine 97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

ACRO448670100EA 3230 DKK
ACRO448670100
5-Bromo-1,3,4-thiadiazol-2-amine 97%
5-Bromo-1,3,4-thiadiazol-2-amine
Formel: C₂H₂BrN₃S
Molekylvægt: 180,02 g/mol
Smeltepunkt: 167…169 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00464325
CAS-nummer: 37566-39-5
UN nummer: 2811
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 165 to 170 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR