4-Methyl-1-naphthaleneboronic acid 96%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
ACRO437530100EA
1990
DKK
ACRO437530100
4-Methyl-1-naphthaleneboronic acid 96%
4-Methyl-1-naphthaleneboronic acid
Formel:
C₁₁H₁₁BO₂ Molekylvægt: 186,02 g/mol Smeltepunkt: 224…228 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01632204 CAS-nummer: 103986-53-4 |
Specifikationer Testresultater
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...