Sodium thiophenolate 90%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
ACRO436260500EA
2060
DKK
ACRO436260500
ACRO436260100
Sodium thiophenolate 90%
Sodium thiophenolate
Formel:
C₆H₅NaS Molekylvægt: 132,1587 g/mol Smeltepunkt: >300 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00066460 CAS-nummer: 930-69-8 UN nummer: 1759 ADR: 8,III |
Specifikationer Testresultater
Appearance | Off-white to yellow to beige Powder |
Infrared spectrum | Conforms |
Titration Iodimetric | ≥88 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...