Butyltrichlorsilan 99%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
ACRO435600250EA
795
DKK
ACRO435600250
Butyltrichlorsilan 99%
Butyltrichlorosilane
Formel:
C₄H₉Cl₃Si Molekylvægt: 191,56 g/mol Kogepunkt: 132…134 °C (1013 hPa) Massefylde: 1,161 g/cm³ Flammepunkt: 54 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00013605 CAS-nummer: 7521-80-4 UN nummer: 1747 ADR: 8,II |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4360 to 1.4380 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...