2-Methyl-2H-indazole-6-boronic acid 97%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
ACRO430490010EA
2520
DKK
ACRO430490010
2-Methyl-2H-indazole-6-boronic acid 97%
2-Methyl-2H-indazole-6-boronic acid
Formel:
C₈H₉BN₂O₂ Molekylvægt: 175,9800 g/mol Smeltepunkt: 306…311 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09870057 CAS-nummer: 1001907-57-8 |
Specifikationer Testresultater
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Specific optical rotation | 3.0° to 5.0° (20°C, 589 nm) (c=1, chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...