Urine sample containers, with screw cap, STERILIN®
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SLIN131BBAC/C50EA 2410 DKK
SLIN131BBAC/C50 SLIN131BBAC/50 SLIN131B SLIN131BBAC SLIN131B/C SLIN131B/50 SLIN131BBAC/C SLIN131B/C50
Urine sample containers, with screw cap, STERILIN®
Containers for use in automated urine analysers.
  • Eliminate the need for decanting in the laboratory
  • Suitable for centrifugation at 3200 ×g
  • Aseptically manufactured
  • A collection cup is also available to simplify transferring the sample into the tube
Certificeringer: Secure screw cap, leak-tested in accordance with BS EN 14254 Annexe D