2-Amino-4'-bromoacetophenone hydrochloride 97%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ACRO427060050EA
819
DKK
ACRO427060050
2-Amino-4'-bromoacetophenone hydrochloride 97%
2-Amino-4'-bromoacetophenone hydrochloride
Formel:
C₈H₉BrClNO Molekylvægt: 250,53 g/mol Smeltepunkt: 280…285 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00051998 CAS-nummer: 5467-72-1 |
Specifikationer Testresultater
Appearance | Beige to pink-orange Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...