(R)-(+)-N,α-Dimethylbenzylamine 99+%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
ACRO395850010EA
640
DKK
ACRO395850010
ACRO395850050
(R)-(+)-N,α-Dimethylbenzylamine 99+%
(R)-(+)-N,α-Dimethylbenzylamine
Formel:
C₉H₁₃N Molekylvægt: 135,2062 g/mol Kogepunkt: 74…76 °C (11 mmHg) Flammepunkt: 61 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00044966 CAS-nummer: 5933-40-4 UN nummer: 2619 ADR: 8,II |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5110 to 1.5130 (20°C, 589 nm) |
GC | ≥99.0 % |
Specific optical rotation | +63° to +67° (20°C, 589 nm) (c=4, methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...