Dowex® 50WX2 (H) 100-200 mesh ion-exchange resin
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
ACRO203031000EA
739
DKK
ACRO203031000
Dowex® 50WX2 (H) 100-200 mesh ion-exchange resin
Dowex® 50WX2 (H)
Opbevaringstemperatur:
Stue temperatur |
MDL Number:
MFCD00132722 CAS-nummer: 12612-37-2 |
Specifikationer Testresultater
Appearance | Beads |
Particle size | ≤15 % on 35 mesh (cumulative) |
Particle size | ≤15 % Through 140 mesh |
Wet volume capacity | ≥0.6 meq/ml H form |
Water retention capacity | 74 to 82 % H form |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...