Phenol flydende/Tris saturated solution stabilised til molekylærbiologi DNase-, RNase-, protease-free

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

ACRO327125000EA 1700 DKK
ACRO327125000
Phenol flydende/Tris saturated solution stabilised til molekylærbiologi DNase-, RNase-, protease-free
Phenol flydende/Tris
Formel: C₆H₅OH
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01632719
CAS-nummer: 108-95-2
UN nummer: 2810
ADR: 6.1,II

Bestil nu

Specifikationer Testresultater

Appearance Yellow to orange Solution
Water 25 to 35 % (K.F.)
Stabilizer 0.08 to 0.12 % hydroxyquinoline
pH 7.8 to 8.2 (at 25°C)
Dnase,Rnase,Protease activity None detected
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR