4-(Diethylamino)butanenitrile 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

ACRO235570010EA 410 DKK
ACRO235570010
4-(Diethylamino)butanenitrile 97%
4-(Diethylamino)butanenitrile

Appearance: Clear yellow to brown Liquid

Formel: C₈H₁₆N₂
Kogepunkt: 89…90 °C (9 mmHg)
Massefylde: 0,84 g/cm³
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 5336-75-4

Bestil nu

Specifikationer Testresultater

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4350 to 1.4370 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR