4(3H)-Pyrimidinone 98+%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

ACRO228250100EA 1300 DKK
ACRO228250100
4(3H)-Pyrimidinone 98+%
4(3H)-Pyrimidinone

Appearance: White to light yellow Powder

Formel: C₄H₄N₂O
Molekylvægt: 96,09 g/mol
Smeltepunkt: 163…168 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00006664
CAS-nummer: 4562-27-0

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 163 to 168 °C
Titration with HClO4 ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR