Anti-V5 Tag Rabbit Polyclonal Antibody (CF488A)
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20440.EA 3290 DKK
20440.
Anti-V5 Tag Rabbit Polyclonal Antibody (CF488A)
Antistoffer
Polyclonal anti-V5 (GKPIPNPLLGLDST) reacts with V5 tagged fusion proteins. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus 5 (SV5). The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. Polyclonal anti-MYC (EQKLISEEDL) reacts with human c- MYC tagged fusion proteins. The antibody recognizes the epitope tag fused to either the amino- or carboxy- termini of targeted proteins. This conjugate is prepared by labeling polyclonal rabbit anti-MYC IgG (H+L) with CF™488A dye. CF™488A (Ex/Em 490/515 nm) is a superior alternative to the traditional green fluorescent dye FITC (or fluorescein) owing to its exceptional brightness and photostability. CF™488A has brightness and photostability comparable to Alexa Fluor® 488.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.
Type: Tag
Antigen: V5 Tag
Clonality: Polyclonal
Clone:
Conjugation: CF488A
Epitope:
Host: Rabbit
Isotype:
Reactivity: MYC tag