Conductivity meter, handheld, CO 301
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
663-0221EA 2350 DKK
VWRI663-0221
Conductivity meter, handheld, CO 301
This device is designed for the measurement of conductivity, resistivity, salinity and TDS in liquids − using a permanently connected electrode (measuring cell). It is suitable for mobile applications and for stationary operation in controlled electromagnetic environments (laboratories).
  • Robust ABS case (IP 65)
  • Min./Max./HOLD function
  • Alarm for low battery and re-calibration
  • Auto-Off function (adjustable: 1 - 120 min.)
  • Clip allows belt attachment, benchtop use and wall-mounting
  • Serial interface (3,5 mm jack) can be connected to the RS232 or USB interface of a PC via an electrically isolated interface converter (not included)
Leveringsinformation: Supplied with conductivity/temperature cell, one standard bottle, case, 9 V battery and manual.